Assay Details
| Gene Symbol: Map3k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011946.3 | NP_036076.2 | 6 - 7 | 907 | 91 | |
| GenBank mRNA | AK143367.1 | - | 6 - 7 | 904 | 91 | |
| U43186.1 | - | 6 - 7 | 805 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k2
Genomic Map