Assay Details
| Gene Symbol: Slc7a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011990.2 | NP_036120.1 | 1 - 2 | 632 | 107 | |
| GenBank mRNA | AB022345.1 | - | 1 - 2 | 599 | 107 | |
| AK037490.1 | - | 1 - 2 | 633 | 107 | ||
| AK037742.1 | - | 1 - 2 | 607 | 107 | ||
| AK076322.1 | - | 1 - 2 | 607 | 107 | ||
| AK089202.1 | - | 1 - 2 | 605 | 107 | ||
| AK162233.1 | - | 1 - 2 | 405 | 107 | ||
| AK163168.1 | - | 1 - 2 | 606 | 107 | ||
| AK165599.1 | - | 1 - 2 | 608 | 107 | ||
| AK171192.1 | - | 1 - 2 | 630 | 107 | ||
| AK171252.1 | - | 1 - 2 | 606 | 107 | ||
| AY766236.1 | - | 1 - 2 | 599 | 107 | ||
| AY766237.1 | - | 599 | 107 | |||
| BC141402.1 | - | 1 - 2 | 529 | 9056015 | 107 | |
| BC141408.1 | - | 1 - 2 | 529 | 9056021 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a11
Genomic Map