Assay Details
| Gene Symbol: Ppp2r5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081457.2 | NP_001074926.1 | 5 - 6 | 748 | 78 | |
| NM_001081458.2 | NP_001074927.1 | 5 - 6 | 748 | 78 | ||
| NM_001135001.1 | NP_001128473.1 | 7 - 8 | 979 | 78 | ||
| NM_012023.3 | NP_036153.2 | 5 - 6 | 748 | 78 | ||
| GenBank mRNA | AB055635.1 | - | 5 - 6 | 598 | 78 | |
| AB055636.1 | - | 5 - 6 | 598 | 78 | ||
| AK031584.1 | - | 5 - 6 | 751 | 78 | ||
| AK129042.1 | - | 7 - 8 | 851 | 78 | ||
| AK190865.1 | - | 89 | 78 | |||
| BC003979.1 | - | 5 - 6 | 708 | 3489999 | 78 | |
| U37353.1 | - | 5 - 6 | 577 | 78 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK012612.1 | - | Chr. - - [-] on Build | 78 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r5c
Genomic Map