Assay Details
| Gene Symbol: Flt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013520.3 | NP_038548.3 | 3 - 4 | 236 | 76 | |
| NR_102725.1 | 3 - 4 | 236 | 76 | |||
| GenBank mRNA | AK020105.1 | - | 3 - 4 | 236 | 76 | |
| BC019801.1 | - | 3 - 4 | 221 | 5132319 | 76 | |
| CK622267.1 | - | 3 - 4 | 206 | 76 | ||
| EU274583.1 | - | 2 - 3 | 148 | 76 | ||
| L23636.1 | - | 2 - 3 | 179 | 76 | ||
| S76459.1 | - | 2 - 3 | 154 | 76 | ||
| U04807.1 | - | 2 - 3 | 403 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Flt3l
Genomic Map