Assay Details
| Gene Symbol: Nr4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013613.2 | NP_038641.1 | 1 - 2 | 317 | 98 | |
| GenBank mRNA | AK158607.1 | - | 1 - 2 | 318 | 98 | |
| BC137715.1 | - | 1 - 2 | 629 | 8860734 | 98 | |
| S53744.1 | - | 1 - 2 | 208 | 98 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK158607.1,BC137715.1,NM_013613.2,S53744.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Transcriptional response to corticotropin-releasing factor in AtT-20 cells.
- Authors
- Peeters PJ, Gohlmann HW, Van den Wyngaert I, Swagemakers SM, Bijnens L, Kass SU, Steckler T
- Journal
- Mol Pharmacol - Page No 1083 (Mon Nov 01 00:00:00 EST 2004)
- Volume (Year)
- 66 (2004)
- Product Usage
- -

Nr4a2
Genomic Map