Assay Details
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244198.1 | NP_001231127.1 | 8 - 9 | 1007 | 97 | |
NM_001244200.1 | NP_001231129.1 | 9 - 10 | 1219 | 97 | ||
NM_001244201.1 | NP_001231130.1 | 7 - 8 | 965 | 97 | ||
NM_001244202.1 | NP_001231131.1 | 7 - 8 | 1521 | 97 | ||
NM_013627.5 | NP_038655.1 | 8 - 9 | 1563 | 97 | ||
GenBank mRNA | AF443223.1 | - | 4 - 5 | 620 | 97 | |
AF457141.1 | - | 2 - 3 | 286 | 97 | ||
AJ292077.1 | - | 7 - 8 | 824 | 97 | ||
AJ307468.1 | - | 7 - 8 | 831 | 97 | ||
AK028059.1 | - | 7 - 8 | 966 | 97 | ||
AK032616.1 | - | 8 - 9 | 1008 | 97 | ||
AK045805.1 | - | 7 - 8 | 1523 | 97 | ||
AK139054.1 | - | 551 | 97 | |||
AY064175.1 | - | 3 - 4 | 430 | 97 | ||
BC011272.1 | - | 8 - 9 | 1251 | 4008490 | 97 | |
BC036957.1 | - | 7 - 8 | 961 | 4504106 | 97 | |
BC069912.1 | - | 4 - 5 | 998 | 97 | ||
M77842.1 | - | 3 - 4 | 550 | 97 | ||
X63963.1 | - | 8 - 9 | 884 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
