Assay Details
| Gene Symbol: Ccl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013652.2 | NP_038680.1 | 1 - 2 | 154 | 70 | |
| GenBank mRNA | AF128218.1 | - | 1 - 2 | 127 | 70 | |
| AF128219.1 | - | 1 - 2 | 127 | 70 | ||
| AK156948.1 | - | 1 - 2 | 147 | 70 | ||
| AK172472.1 | - | 1 - 2 | 147 | 70 | ||
| BC119257.1 | - | 1 - 2 | 101 | 8734006 | 70 | |
| BC119259.1 | - | 1 - 2 | 101 | 8734008 | 70 | |
| BY163868.1 | - | 1 - 2 | 155 | 70 | ||
| M23503.1 | - | 1 - 2 | 152 | 70 | ||
| M35590.1 | - | 1 - 2 | 137 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl4
Genomic Map