Assay Details
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013657.5 | NP_038685.3 | 1 - 2 | 150 | 80 | |
GenBank mRNA | AK034239.1 | - | 1 - 2 | 209 | 80 | |
AK078589.1 | - | 1 - 2 | 202 | 80 | ||
AK140348.1 | - | 686 | 80 | |||
BC066852.1 | - | 1 - 2 | 150 | 30093230 | 80 | |
X85994.1 | - | 1 - 2 | 102 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK034239.1,AK140348.1,BC066852.1,NM_013657.5,X85994.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
