Assay Details
| Gene Symbol: Tap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013683.2 | NP_038711.2 | 6 - 7 | 1630 | 77 | |
| GenBank mRNA | AK151481.1 | - | 6 - 7 | 1385 | 77 | |
| AK153890.1 | - | 6 - 7 | 1634 | 77 | ||
| AK162731.1 | - | 6 - 7 | 1430 | 77 | ||
| AK166046.1 | - | 6 - 7 | 1631 | 77 | ||
| AK167485.1 | - | 6 - 7 | 1632 | 77 | ||
| BC013802.1 | - | 6 - 7 | 1041 | 3257566 | 77 | |
| BC024897.1 | - | 6 - 7 | 1153 | 3257734 | 77 | |
| BC031584.1 | - | 2 - 3 | 1765 | 77 | ||
| KJ650227.1 | - | 6 - 7 | 1306 | 77 | ||
| M55637.1 | - | 6 - 7 | 1022 | 77 | ||
| U60018.1 | - | 6 - 7 | 1306 | 77 | ||
| U60019.1 | - | 6 - 7 | 1306 | 77 | ||
| U60020.1 | - | 6 - 7 | 1306 | 77 | ||
| U60021.1 | - | 6 - 7 | 1306 | 77 | ||
| U60023.1 | - | 6 - 7 | 1306 | 77 | ||
| X59615.1 | - | 6 - 7 | 865 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tap1
Genomic Map