Assay Details
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161420.1 | NP_001154892.1 | 6 - 7 | 1540 | 117 | |
| NM_013703.2 | NP_038731.2 | 6 - 7 | 1540 | 117 | ||
| GenBank mRNA | AK136838.1 | - | 6 - 7 | 1360 | 117 | |
| L33417.1 | - | 6 - 7 | 964 | 117 | ||
| U06670.1 | - | 6 - 7 | 1126 | 117 | ||
More Information
Set Membership: |
|

Vldlr
Genomic Map