Assay Details
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161420.1 | NP_001154892.1 | 14 - 15 | 2694 | 102 | |
| NM_013703.2 | NP_038731.2 | 14 - 15 | 2694 | 102 | ||
| GenBank mRNA | AK044683.1 | - | 6 - 7 | 1302 | 102 | |
| AK076172.1 | - | 14 - 15 | 2699 | 102 | ||
| AK136838.1 | - | 14 - 15 | 2514 | 102 | ||
| BC013622.1 | - | 14 - 15 | 3968213 | 102 | ||
| L33417.1 | - | 14 - 15 | 2118 | 102 | ||
| U06670.1 | - | 14 - 15 | 2280 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vldlr
Genomic Map