Assay Details
| Gene Symbol: Pdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013743.2 | NP_038771.1 | 7 - 8 | 888 | 72 | |
| GenBank mRNA | AJ001418.1 | - | 7 - 8 | 888 | 72 | |
| AK004543.1 | - | 7 - 8 | 913 | 72 | ||
| AK036818.1 | - | 7 - 8 | 920 | 72 | ||
| BC026134.1 | - | 7 - 8 | 917 | 4188580 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdk4
Genomic Map