Assay Details
Gene Symbol: Csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198841.1 | NP_001185770.1 | 1 - 2 | 60 | 62 | |
GenBank mRNA | AK003120.1 | - | 1 - 2 | 44 | 62 | |
BG793105.1 | - | 60 | 62 | |||
BY067937.1 | - | 1 - 2 | 40 | 62 | ||
Z49883.1 | - | 1 - 2 | 45 | 62 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK003120.1,NM_001198841.1,Z49883.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
