Assay Details
Gene Symbol: Csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198841.1 | NP_001185770.1 | 3 - 4 | 362 | 59 | |
NM_013808.4 | NP_038836.1 | 3 - 4 | 384 | 59 | ||
GenBank mRNA | AI314553.1 | - | 3 - 4 | 384 | 59 | |
AK003120.1 | - | 3 - 4 | 346 | 59 | ||
AK011624.1 | - | 3 - 4 | 341 | 59 | ||
BC061131.1 | - | 3 - 4 | 364 | 30306316 | 59 | |
BY067937.1 | - | 3 - 4 | 342 | 59 | ||
D88791.1 | - | 2 - 3 | 282 | 59 | ||
Z49883.1 | - | 3 - 4 | 347 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
