Assay Details
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013820.3 | NP_038848.1 | 10 - 11 | 2039 | 70 | |
GenBank mRNA | AJ238540.1 | - | 10 - 11 | 1982 | 70 | |
AK145936.1 | - | 10 - 11 | 2020 | 70 | ||
BC054472.1 | - | 10 - 11 | 1958 | 6465367 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
