Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300012589.mm.1 |
NM_013846.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012589.mm.1 |
NM_013846.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012589.mm.1 |
TC1300002015.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300012591.mm.1 |
NM_013846.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012591.mm.1 |
NM_013846.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012591.mm.1 |
TC1300002015.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300012594.mm.1 |
NM_013846.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012594.mm.1 |
NM_013846.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012594.mm.1 |
TC1300002015.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300012590.mm.1 |
NM_013846.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012590.mm.1 |
NM_013846.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012590.mm.1 |
TC1300002015.mm.1 |
MTA TC |