Assay Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145959.1 | NP_001139431.1 | 6 - 7 | 597 | 70 | |
| NM_013864.2 | NP_038892.1 | 7 - 8 | 639 | 70 | ||
| GenBank mRNA | AB033921.1 | - | 7 - 8 | 502 | 70 | |
| AK076514.1 | - | 4 - 5 | 273 | 70 | ||
| AK154597.1 | - | 7 - 8 | 507 | 70 | ||
| AK158900.1 | - | 6 - 7 | 506 | 70 | ||
| AK168011.1 | - | 6 - 7 | 464 | 70 | ||
| AK173136.1 | - | 4 - 5 | 280 | 70 | ||
| BC012963.1 | - | 7 - 8 | 506 | 4211421 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndrg2
Genomic Map