Assay Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145959.1 | NP_001139431.1 | 13 - 14 | 1027 | 78 | |
| NM_013864.2 | NP_038892.1 | 14 - 15 | 1069 | 78 | ||
| GenBank mRNA | AB033921.1 | - | 14 - 15 | 932 | 78 | |
| AK076514.1 | - | 11 - 12 | 703 | 78 | ||
| AK154597.1 | - | 14 - 15 | 937 | 78 | ||
| AK158900.1 | - | 13 - 14 | 936 | 78 | ||
| AK168011.1 | - | 13 - 14 | 894 | 78 | ||
| AK173136.1 | - | 11 - 12 | 710 | 78 | ||
| BC012963.1 | - | 14 - 15 | 936 | 4211421 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndrg2
Genomic Map