Assay Details
| Gene Symbol: Timm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013899.2 | NP_038927.2 | 2 - 3 | 243 | 86 | |
| GenBank mRNA | AF150090.1 | - | 2 - 3 | 220 | 86 | |
| AK008088.1 | - | 2 - 3 | 244 | 86 | ||
| BC031448.1 | - | 2 - 3 | 221 | 3257567 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394887 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394888 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394881 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957003 | BC031448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957003 | NM_013899.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957003 | AF150090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957003 | AK008088.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957004 | NM_013899.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957004 | AF150090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957004 | AK008088.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899151 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166791 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957004 | NM_013899.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957004 | AF150090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957004 | AK008088.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957003 | BC031448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957003 | NM_013899.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957003 | AF150090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957003 | AK008088.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348624 | BC031448.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348624 | NM_013899.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348624 | AF150090.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348624 | AK008088.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394818 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166791 | - | - |
| Whole Mouse Genome G4122A | A_51_P348624 | BC031448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P348624 | NM_013899.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P348624 | AF150090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P348624 | AK008088.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127780 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166791 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02173329 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394887 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21394888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02173324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02173325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02173326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02272500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21394825 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10473366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473365 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417499_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473363 | - | - |
| Mouse Genome 430 2.0 Array | 1417499_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417499_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30059_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30059_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200009760.mm.1 | NM_013899.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200009760.mm.1 | NM_013899.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200009760.mm.1 | TC0200001223.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200009759.mm.1 | NM_013899.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200009759.mm.1 | NM_013899.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200009759.mm.1 | TC0200001223.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417499_at | - | - |
| Mouse Expression Set 430 | 1455211_a_at | - | - |
| Mouse Expression Set 430 | 1455211_PM_a_at | - | - |
| Mouse Expression Set 430 | 1417499_PM_at | - | - |
| Murine Genome U74v2 Set | 94012_at | - | - |
| Murine Genome U74v2 Set | 94210_at | - | - |
| Murine Genome U74v2 Set | 94012_PM_at | - | - |
| Murine Genome U74v2 Set | 94210_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417499_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417499_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455211_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455211_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 587499 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2450601 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2450601 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002450601 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227235 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227235 | - | - |
| mouse-6 v1.1 (Illumina) | scl20689.4.1_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2450601 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20689.4.1_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2450601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1227235 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227235 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20689.4.1_4-S | - | - |

Timm10
Genomic Map