Assay Details
| Gene Symbol: Fbxw4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013907.2 | NP_038935.1 | 5 - 6 | 929 | 73 | |
| GenBank mRNA | AB022163.1 | - | 5 - 6 | 929 | 73 | |
| BC027031.1 | - | 5 - 6 | 791 | 5364017 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxw4
Genomic Map