Assay Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271898.1 | NP_001258827.1 | 3 - 4 | 774 | 66 | |
| GenBank mRNA | AB034914.1 | - | 3 - 4 | 463 | 66 | |
| AF006688.1 | - | 3 - 4 | 513 | 66 | ||
| AK128941.1 | - | 3 - 4 | 476 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acox1
Genomic Map