Assay Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271898.1 | NP_001258827.1 | 5 - 6 | 1001 | 101 | |
| NM_015729.3 | NP_056544.2 | 5 - 6 | 1001 | 101 | ||
| GenBank mRNA | AB034914.1 | - | 5 - 6 | 690 | 101 | |
| AF006688.1 | - | 5 - 6 | 740 | 101 | ||
| AK034760.1 | - | 2 - 3 | 476 | 101 | ||
| AK040566.1 | - | 5 - 6 | 704 | 101 | ||
| AK054446.1 | - | 5 - 6 | 777 | 101 | ||
| AK128941.1 | - | 5 - 6 | 703 | 101 | ||
| AK170217.1 | - | 1 - 2 | 71 | 101 | ||
| BC054727.1 | - | 5 - 6 | 669 | 101 | ||
| BC056448.1 | - | 5 - 6 | 1001 | 5704873 | 101 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK170217.1 transcripts.
Set Membership: |
|

Acox1
Genomic Map