Assay Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271898.1 | NP_001258827.1 | 8 - 9 | 1445 | 74 | |
| NM_015729.3 | NP_056544.2 | 8 - 9 | 1445 | 74 | ||
| GenBank mRNA | AB034914.1 | - | 8 - 9 | 1134 | 74 | |
| AF006688.1 | - | 8 - 9 | 1184 | 74 | ||
| AK034760.1 | - | 4 - 5 | 750 | 74 | ||
| AK040566.1 | - | 8 - 9 | 1148 | 74 | ||
| AK054446.1 | - | 8 - 9 | 1221 | 74 | ||
| AK128941.1 | - | 8 - 9 | 1147 | 74 | ||
| AK170217.1 | - | 4 - 5 | 515 | 74 | ||
| BC054727.1 | - | 8 - 9 | 1113 | 74 | ||
| BC056448.1 | - | 8 - 9 | 1445 | 5704873 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox1
Genomic Map