Assay Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271898.1 | NP_001258827.1 | 10 - 11 | 1824 | 71 | |
| NM_015729.3 | NP_056544.2 | 10 - 11 | 1824 | 71 | ||
| GenBank mRNA | AB034914.1 | - | 10 - 11 | 1513 | 71 | |
| AF006688.1 | - | 10 - 11 | 1563 | 71 | ||
| AK034760.1 | - | 6 - 7 | 1129 | 71 | ||
| AK040566.1 | - | 10 - 11 | 1527 | 71 | ||
| AK128941.1 | - | 10 - 11 | 1526 | 71 | ||
| AK170217.1 | - | 6 - 7 | 894 | 71 | ||
| BC054727.1 | - | 10 - 11 | 1492 | 71 | ||
| BC056448.1 | - | 10 - 11 | 1824 | 5704873 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox1
Genomic Map