Assay Details
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042513.1 | NP_001035978.1 | 5 - 6 | 517 | 69 | |
| NM_001042514.1 | NP_001035979.1 | 4 - 5 | 605 | 69 | ||
| NM_001042523.1 | NP_001035988.1 | 5 - 6 | 734 | 69 | ||
| NM_015762.2 | NP_056577.2 | 4 - 5 | 410 | 69 | ||
| GenBank mRNA | AB027565.1 | - | 4 - 5 | 437 | 69 | |
| AF333036.1 | - | 5 - 6 | 734 | 69 | ||
| AK011902.1 | - | 5 - 6 | 518 | 69 | ||
| AK146125.1 | - | 4 - 5 | 411 | 69 | ||
| AK149625.1 | - | 5 - 6 | 518 | 69 | ||
| AK151927.1 | - | 5 - 6 | 518 | 69 | ||
| AK168356.1 | - | 5 - 6 | 518 | 69 | ||
| AK172383.1 | - | 4 - 5 | 443 | 69 | ||
| AK191660.1 | - | 1 - 2 | 107 | 69 | ||
| AK211217.1 | - | 1 - 2 | 107 | 69 | ||
| AK212800.1 | - | 1 - 2 | 107 | 69 | ||
| BC020274.1 | - | 4 - 5 | 372 | 69 | ||
| BC037643.1 | - | 4 - 5 | 690 | 5059831 | 69 | |
| CJ136679.1 | - | 5 - 6 | 518 | 69 | ||
| CX562215.1 | - | 4 - 5 | 605 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd1
Genomic Map