Assay Details
| Gene Symbol: Tfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289507.1 | NP_001276436.1 | 2 - 3 | 487 | 62 | |
| NM_001289509.1 | NP_001276438.1 | 4 - 5 | 506 | 62 | ||
| NM_001289511.1 | NP_001276440.1 | 3 - 4 | 364 | 62 | ||
| NM_015799.4 | NP_056614.3 | 4 - 5 | 446 | 62 | ||
| GenBank mRNA | AF207741.1 | - | 2 - 3 | 321 | 62 | |
| AF222895.1 | - | 3 - 4 | 364 | 62 | ||
| AK004848.2 | - | 4 - 5 | 422 | 62 | ||
| AK004965.1 | - | 4 - 5 | 477 | 62 | ||
| AK155526.1 | - | 3 - 4 | 584 | 62 | ||
| AK165320.1 | - | 2 - 3 | 315 | 62 | ||
| BC013654.1 | - | 2 - 3 | 487 | 4196597 | 62 | |
More Information
Set Membership: |
|

Tfr2
Genomic Map