Assay Details
| Gene Symbol: Tfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289507.1 | NP_001276436.1 | 10 - 11 | 1584 | 93 | |
| NM_001289509.1 | NP_001276438.1 | 12 - 13 | 1603 | 93 | ||
| NM_001289511.1 | NP_001276440.1 | 11 - 12 | 1461 | 93 | ||
| NM_015799.4 | NP_056614.3 | 12 - 13 | 1543 | 93 | ||
| GenBank mRNA | AF207741.1 | - | 10 - 11 | 1418 | 93 | |
| AF222895.1 | - | 11 - 12 | 1461 | 93 | ||
| AK004848.2 | - | 12 - 13 | 1519 | 93 | ||
| AK004965.1 | - | 12 - 13 | 1574 | 93 | ||
| AK155526.1 | - | 11 - 12 | 1681 | 93 | ||
| AK165320.1 | - | 10 - 11 | 1412 | 93 | ||
| BC013654.1 | - | 10 - 11 | 1584 | 4196597 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126441 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126440 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955532 | AK155526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955532 | BB736789.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955532 | AK004965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955532 | AK004848.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955532 | NM_015799.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668309 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 857067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42960 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110484 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177581 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955532 | AK155526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955532 | BB736789.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955532 | AK004965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955532 | AK004848.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955532 | NM_015799.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101474 | BC013654.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101474 | AK165320.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101474 | AK155526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101474 | AK004965.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101474 | AK004848.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126411 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05760935 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110484 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177581 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05760952 | - | - |
| Whole Mouse Genome G4122A | A_51_P101474 | BC013654.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P101474 | AK165320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P101474 | AK155526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P101474 | AK004965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P101474 | AK004848.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118369 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110484 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04728143 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04728136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04546147 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04728142 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04546160 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04546156 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04728137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126441 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04550141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04683301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04728145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04316418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04546153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04546147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04232323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04145343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04712580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04728135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04683306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04728142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04728136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04232328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04683300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04777052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126446 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526648 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526647 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526631 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526649 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526646 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526632 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459994_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425381_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526630 | - | - |
| Mouse Genome 430 2.0 Array | 1425381_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425381_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1459994_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1459994_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50765_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50765_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500014811.mm.1 | TC0500001629.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014811.mm.1 | NM_001289507.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014811.mm.1 | NM_001289509.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014811.mm.1 | NM_001289511.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014811.mm.1 | NM_015799.4 | Refseq |
| Mouse Expression Set 430 | 1459994_x_at | - | - |
| Mouse Expression Set 430 | 1425381_a_at | - | - |
| Mouse Expression Set 430 | 1425381_PM_a_at | - | - |
| Mouse Expression Set 430 | 1459994_PM_x_at | - | - |
| Murine Genome U74v2 Set | 160674_PM_at | - | - |
| Murine Genome U74v2 Set | 160674_at | - | - |
| Murine Genome U74v2 Set | 137177_at | - | - |
| Murine Genome U74v2 Set | 170121_at | - | - |
| Murine Genome U74v2 Set | 170121_PM_at | - | - |
| Murine Genome U74v2 Set | 137177_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425381_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1459994_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1459994_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425381_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1459994_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425381_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425381_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1459994_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 923412 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4760397 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5290070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5290070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005290070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760397 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2450491 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227831 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227831 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2450491 | - | - |
| mouse-6 v1.1 (Illumina) | scl0004096.1_49-S | - | - |
| mouse-6 v1.1 (Illumina) | scl27100.19.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5290070 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27100.19.1_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004096.1_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760397 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1227831 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2450491 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227831 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2450491 | - | - |

Tfr2
Genomic Map