Assay Details
| Gene Symbol: Atp11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293667.1 | NP_001280596.1 | 6 - 7 | 864 | 79 | |
| NM_001293668.1 | NP_001280597.1 | 6 - 7 | 864 | 79 | ||
| NM_015804.3 | NP_056619.1 | 6 - 7 | 864 | 79 | ||
| NR_121603.1 | 6 - 7 | 864 | 79 | |||
| GenBank mRNA | AF156551.1 | - | 6 - 7 | 656 | 79 | |
| AK147497.1 | - | 6 - 7 | 867 | 79 | ||
| AK173088.1 | - | 6 - 7 | 811 | 79 | ||
| BC138715.1 | - | 6 - 7 | 583 | 8861737 | 79 | |
| BC145399.1 | - | 6 - 7 | 584 | 79 | ||
| BC145400.1 | - | 6 - 7 | 584 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp11a
Genomic Map