Assay Details
| Gene Symbol: Ccnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122982.2 | NP_001116454.1 | 2 - 3 | 324 | 126 | |
| NM_001290420.1 | NP_001277349.1 | 2 - 3 | 324 | 126 | ||
| NM_001290422.1 | NP_001277351.1 | 2 - 3 | 324 | 126 | ||
| NM_016746.4 | NP_058026.2 | 2 - 3 | 324 | 126 | ||
| GenBank mRNA | AK009615.1 | - | 2 - 3 | 269 | 126 | |
| AK035728.1 | - | 2 - 3 | 269 | 126 | ||
| AK035762.1 | - | 2 - 3 | 285 | 126 | ||
| AK037957.1 | - | 2 - 3 | 325 | 126 | ||
| AK135472.1 | - | 2 - 3 | 320 | 126 | ||
| BB631264.1 | - | 2 - 3 | 322 | 126 | ||
| BC003344.1 | - | 2 - 3 | 257 | 3257730 | 126 | |
| BC029211.1 | - | 2 - 3 | 270 | 4526925 | 126 | |
| BC037689.1 | - | 2 - 3 | 252 | 126 | ||
| BC062376.1 | - | 2 - 3 | 239 | 126 | ||
| BC087544.1 | - | 2 - 3 | 256 | 126 | ||
| BC100396.1 | - | 2 - 3 | 298 | 126 | ||
| BC120649.1 | - | 2 - 3 | 251 | 40129572 | 126 | |
| BC120677.1 | - | 2 - 3 | 251 | 40129600 | 126 | |
| U62638.1 | - | 2 - 3 | 299 | 126 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC029211.1 transcripts.
Set Membership: |
|

Ccnc
Genomic Map