Assay Details
| Gene Symbol: Rgs14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016758.3 | NP_058038.2 | 11 - 12 | 1420 | 78 | |
| GenBank mRNA | AK002891.1 | - | 6 - 7 | 909 | 78 | |
| BC030321.1 | - | 11 - 12 | 1392 | 5374531 | 78 | |
| U85055.1 | - | 11 - 12 | 1260 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs14
Genomic Map