Assay Details
Gene Symbol: Itgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016780.2 | NP_058060.2 | 2 - 3 | 241 | 83 | |
GenBank mRNA | AF026509.1 | - | 2 - 3 | 185 | 83 | |
AK157958.1 | - | 2 - 3 | 187 | 83 | ||
AY541673.1 | - | 2 - 3 | 155 | 83 | ||
BC125518.1 | - | 2 - 3 | 241 | 40130033 | 83 | |
BC125520.1 | - | 2 - 3 | 241 | 40130035 | 83 | |
U83167.1 | - | 2 - 3 | 152 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
