Assay Details
| Gene Symbol: Pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016783.3 | NP_058063.2 | 1 - 2 | 392 | 64 | |
| GenBank mRNA | AF042491.1 | - | 1 - 2 | 343 | 64 | |
| AK153272.1 | - | 1 - 2 | 409 | 64 | ||
| AK159101.1 | - | 1 - 2 | 409 | 64 | ||
| AK159345.1 | - | 1 - 2 | 409 | 64 | ||
| AK159599.1 | - | 1 - 2 | 409 | 64 | ||
| AK162315.1 | - | 1 - 2 | 412 | 64 | ||
| AK166240.1 | - | 1 - 2 | 409 | 64 | ||
| AK167655.1 | - | 1 - 2 | 409 | 64 | ||
| AK168387.1 | - | 1 - 2 | 408 | 64 | ||
| AK168882.1 | - | 1 - 2 | 409 | 64 | ||
| AK169063.1 | - | 2 - 3 | 352 | 64 | ||
| BC006016.1 | - | 1 - 2 | 392 | 3591536 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319267 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319274 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319269 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319266 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319268 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319272 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144581 | AF042491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144581 | AF042491.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161882 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139351 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144581 | AF042491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144581 | AF042491.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32319254 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139351 | - | - |
| Whole Mouse Genome G4122A | A_51_P144581 | AF042491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144581 | AF042491.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127191 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14637339 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14637327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14637332 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319274 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319272 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319266 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319268 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319269 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32319267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32319230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15189701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14544267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15069696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14770330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15069691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15099175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15069694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15191041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14640591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15150991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14640605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15189702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14578137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14790973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32775493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14770327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14637346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14637338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32319305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10599203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599202 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423451_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10599200 | - | - |
| Mouse Genome 430 2.0 Array | 1423451_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423451_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53328_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53328_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00002166.mm.1 | NM_016783.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002166.mm.1 | TC0X00000323.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00002165.mm.1 | NM_016783.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002165.mm.1 | TC0X00000323.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00002167.mm.1 | NM_016783.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002167.mm.1 | TC0X00000323.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423451_PM_at | - | - |
| Mouse Expression Set 430 | 1423451_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423451_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423451_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423451_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423451_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 577557 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4150692 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4150692 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004150692 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2623216 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2623216 | - | - |
| mouse-6 v1.1 (Illumina) | scl54996.3_58-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4150692 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54996.3_58-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150692 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2623216 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2623216 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl54996.3_58-S | - | - |

Pgrmc1
Genomic Map