Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 15 - 16 | 3341 | 93 | |
| NM_016813.2 | NP_058093.2 | 16 - 17 | 1590 | 93 | ||
| GenBank mRNA | AF093140.1 | - | 16 - 17 | 1566 | 93 | |
| AK052937.1 | - | 6 - 7 | 757 | 93 | ||
| AK077644.1 | - | 16 - 17 | 1547 | 93 | ||
| AK079583.1 | - | 16 - 17 | 1579 | 93 | ||
| AK086527.1 | - | 15 - 16 | 3352 | 93 | ||
| AK133883.1 | - | 16 - 17 | 1579 | 93 | ||
| AK143640.1 | - | 3342 | 93 | |||
| AK167518.1 | - | 16 - 17 | 1578 | 93 | ||
| BC005594.1 | - | 16 - 17 | 1590 | 3710546 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map