Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
NM_001253682.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
NM_001253682.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
NM_001253683.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
NM_001253683.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
NM_016984.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
NM_016984.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
NR_046161.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
NR_046161.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002788.mm.1 |
TC0300000407.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
NM_001253682.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
NM_001253682.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
NM_001253683.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
NM_001253683.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
NM_016984.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
NM_016984.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
NR_046161.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
NR_046161.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002784.mm.1 |
TC0300000407.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
NM_001253682.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
NM_001253682.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
NM_001253683.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
NM_001253683.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
NM_016984.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
NM_016984.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
NR_046161.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
NR_046161.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002785.mm.1 |
TC0300000407.mm.1 |
MTA TC |