Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100028405.mm.1 |
NM_017366.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028405.mm.1 |
TC1100003083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028403.mm.1 |
NM_017366.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028403.mm.1 |
TC1100003083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028406.mm.1 |
NM_017366.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028406.mm.1 |
TC1100003083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028407.mm.1 |
NM_017366.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028407.mm.1 |
TC1100003083.mm.1 |
MTA TC |