Assay Details
| Gene Symbol: Atp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290627.1 | NP_001277556.1 | 6 - 7 | 830 | 85 | |
| NM_018731.3 | NP_061201.2 | 6 - 7 | 830 | 85 | ||
| GenBank mRNA | AK143905.1 | - | 6 - 7 | 831 | 85 | |
| BC015262.1 | - | 6 - 7 | 816 | 4241977 | 85 | |
| U17282.1 | - | 5 - 6 | 797 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp4a
Genomic Map