Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900015624.mm.1 |
NM_001205312.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015624.mm.1 |
NM_001205312.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015624.mm.1 |
NM_018793.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015624.mm.1 |
NM_018793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015624.mm.1 |
TC0900001858.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900015623.mm.1 |
NM_001205312.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015623.mm.1 |
NM_001205312.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015623.mm.1 |
NM_018793.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015623.mm.1 |
NM_018793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015623.mm.1 |
TC0900001858.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900015625.mm.1 |
NM_001205312.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015625.mm.1 |
NM_001205312.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015625.mm.1 |
NM_018793.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015625.mm.1 |
NM_018793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015625.mm.1 |
TC0900001858.mm.1 |
MTA TC |