Assay Details
| Gene Symbol: Syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018803.2 | NP_061273.1 | 4 - 5 | 1286 | 69 | |
| GenBank mRNA | AB026807.1 | - | 4 - 5 | 1195 | 69 | |
| BC125632.1 | - | 4 - 5 | 1228 | 40130147 | 69 | |
| BC125634.1 | - | 4 - 5 | 1232 | 40130149 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syt10
Genomic Map