Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200013399.mm.1 |
NM_019392.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013399.mm.1 |
NM_019392.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013399.mm.1 |
TC0200001781.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013402.mm.1 |
NM_019392.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013402.mm.1 |
NM_019392.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013402.mm.1 |
TC0200001781.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013401.mm.1 |
NM_019392.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013401.mm.1 |
NM_019392.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013401.mm.1 |
TC0200001781.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013398.mm.1 |
NM_019392.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013398.mm.1 |
NM_019392.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013398.mm.1 |
TC0200001781.mm.1 |
MTA TC |