Assay Details
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290800.1 | NP_001277729.1 | 14 - 15 | 2465 | 100 | |
NM_019392.2 | NP_062265.2 | 14 - 15 | 2001 | 100 | ||
GenBank mRNA | AB000828.1 | - | 14 - 15 | 1738 | 100 | |
AK141198.1 | - | 14 - 15 | 1772 | 100 | ||
BC066058.1 | - | 14 - 15 | 2001 | 6834015 | 100 | |
BC082325.1 | - | 14 - 15 | 1949 | 30356080 | 100 | |
D17393.1 | - | 14 - 15 | 2443 | 100 | ||
U05683.1 | - | 14 - 15 | 1786 | 100 | ||
U18342.1 | - | 14 - 15 | 1918 | 100 | ||
U18343.1 | - | 14 - 15 | 2461 | 100 | ||
U18933.1 | - | 14 - 15 | 1937 | 100 | ||
X78103.1 | - | 14 - 15 | 1945 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
