Assay Details
| Gene Symbol: Ndufa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019443.2 | NP_062316.1 | 1 - 2 | 250 | 89 | |
| GenBank mRNA | AI844651.1 | - | 2 - 3 | 89 | ||
| AK007840.1 | - | 1 - 2 | 157 | 89 | ||
| AK131584.1 | - | 449 | 89 | |||
| BC024364.1 | - | 1 - 2 | 159 | 5031206 | 89 | |
| CT010398.1 | - | 1 - 2 | 102 | 89 | ||
| Y07708.1 | - | 1 - 2 | 269 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322119 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322122 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | NM_019443.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | AI844651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | AK007840.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | BC024364.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | CF383819.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | NM_019443.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | AI844651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | AK007840.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | BC024364.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139747 | CF383819.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939983 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19381 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128483 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | NM_019443.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | AI844651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | AK007840.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | BC024364.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | CF383819.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | NM_019443.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | AI844651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | AK007840.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | BC024364.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139747 | CF383819.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | NM_019443.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | AI844651.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | AK007840.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | BC024364.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | CF383819.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | NM_019443.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | AI844651.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | AK007840.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | BC024364.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139747 | CF383819.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P472405 | NM_019443.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P472405 | AI844651.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P472405 | AK007840.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P472405 | BC024364.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P472405 | CF383819.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322105 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128483 | - | - |
| Whole Mouse Genome G4122A | A_52_P139747 | NM_019443.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | AI844651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | AK007840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | BC024364.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | CF383819.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | NM_019443.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | AI844651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | AK007840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | BC024364.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139747 | CF383819.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472405 | NM_019443.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472405 | AI844651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472405 | AK007840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472405 | BC024364.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472405 | CF383819.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109478 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128483 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14748050 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14555201 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14555209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14555202 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14682220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14512763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14793142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14748049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14512764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32322113 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10604102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604105 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422241_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604100 | - | - |
| Mouse Genome 430 2.0 Array | 1422241_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422241_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54405_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54405_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00013116.mm.1 | NM_019443.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013116.mm.1 | NM_019443.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013116.mm.1 | TC0X00002061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013115.mm.1 | NM_019443.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013115.mm.1 | NM_019443.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013115.mm.1 | TC0X00002061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013118.mm.1 | NM_019443.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013118.mm.1 | NM_019443.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013118.mm.1 | TC0X00002061.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422241_a_at | - | - |
| Mouse Expression Set 430 | 1422241_PM_a_at | - | - |
| Murine Genome U74v2 Set | 98118_at | - | - |
| Murine Genome U74v2 Set | 98118_PM_at | - | - |
| Murine Genome U74v2 Set | 98117_at | - | - |
| Murine Genome U74v2 Set | 98117_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422241_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422241_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422241_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422241_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 555568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6200411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200411 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251370 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251370 | - | - |
| mouse-6 v1.1 (Illumina) | scl54331.2.4_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200411 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54331.2.4_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200411 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1251370 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251370 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl54331.2.4_4-S | - | - |

Ndufa1
Genomic Map