Assay Details
| Gene Symbol: Asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019652.1 | NP_062626.1 | 3 - 4 | 466 | 65 | |
| GenBank mRNA | AF039405.2 | - | 3 - 4 | 466 | 65 | |
| AK007827.1 | - | 3 - 4 | 469 | 65 | ||
| AK171690.1 | - | 3 - 4 | 470 | 65 | ||
| AK208780.1 | - | 2 - 3 | 261 | 65 | ||
| AK216123.1 | - | 2 - 3 | 261 | 65 | ||
| BC016453.1 | - | 3 - 4 | 454 | 4920063 | 65 | |
| BC018430.1 | - | 1 - 2 | 94 | 2616359 | 65 | |
| BC083335.1 | - | 3 - 4 | 464 | 5717763 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023267 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P469531 | BC016453.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P469531 | AK171690.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P469531 | NM_019652.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P469531 | AF039405.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P469531 | BC016453.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P469531 | AK171690.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P469531 | NM_019652.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P469531 | AF039405.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355583 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174549 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178425 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P469531 | BC016453.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P469531 | AK171690.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P469531 | NM_019652.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P469531 | AF039405.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P469531 | BC016453.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P469531 | AK171690.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P469531 | NM_019652.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P469531 | AF039405.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469531 | BC016453.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469531 | AK171690.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469531 | NM_019652.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469531 | AF039405.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469531 | BC016453.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469531 | AK171690.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469531 | NM_019652.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469531 | AF039405.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023244 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01784530 | - | - |
| Whole Mouse Genome G4122A | A_51_P469531 | BC016453.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469531 | AK171690.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469531 | NM_019652.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469531 | AF039405.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469531 | BC016453.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469531 | AK171690.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469531 | NM_019652.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469531 | AF039405.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106019 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135433 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178425 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07141463 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07141467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07140099 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07141460 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07141459 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07140102 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07141481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07141478 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07140094 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07141474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26023267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07141475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07079574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07086705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07141459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07376389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07079569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06917665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07141478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07235966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07270964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07218126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023210 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10580304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580306 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418292_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10580300 | - | - |
| Mouse Genome 430 2.0 Array | 1418292_at | - | - |
| Mouse Genome 430 2.0 Array | 1418292_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56495_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56495_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800019964.mm.1 | NM_019652.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019964.mm.1 | NM_019652.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019964.mm.1 | TC0800002599.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418292_at | - | - |
| Mouse Expression Set 430 | 1418292_PM_at | - | - |
| Murine Genome U74v2 Set | 92428_at | - | - |
| Murine Genome U74v2 Set | 92428_PM_at | - | - |
| Murine Genome U74v2 Set | 169775_r_at | - | - |
| Murine Genome U74v2 Set | 169775_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418292_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418292_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418292_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418292_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3830041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3830041 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003830041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2683374 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2683374 | - | - |
| mouse-6 v1.1 (Illumina) | scl34546.7.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3830041 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34546.7.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830041 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2683374 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2683374 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34546.7.1_7-S | - | - |

Asna1
Genomic Map