Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016205.mm.1 |
NM_001164107.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016205.mm.1 |
NM_001164107.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016205.mm.1 |
NM_001164108.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016205.mm.1 |
NM_001164108.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016205.mm.1 |
NM_019955.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016205.mm.1 |
NM_019955.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016205.mm.1 |
TC1400002103.mm.1 |
MTA TC |