Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200011192.mm.1 |
NM_001146200.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011192.mm.1 |
NM_001146200.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011192.mm.1 |
NM_001146201.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011192.mm.1 |
NM_001146201.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011192.mm.1 |
NM_020272.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011192.mm.1 |
NM_020272.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011192.mm.1 |
TC1200001605.mm.1 |
MTA TC |