Assay Details
Gene Symbol: Retnla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020509.3 | NP_065255.2 | 3 - 4 | 313 | 86 | |
GenBank mRNA | AF205951.1 | - | 2 - 3 | 246 | 86 | |
AF290872.1 | - | 3 - 4 | 351 | 86 | ||
AF316397.2 | - | 3 - 4 | 255 | 86 | ||
AF323082.1 | - | 3 - 4 | 323 | 86 | ||
AK007553.1 | - | 3 - 4 | 258 | 86 | ||
BC029248.1 | - | 2 - 3 | 253 | 4189285 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | AK007553.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | BC029248.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | NM_020509.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | AF205951.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | AF290872.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | AK007553.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | BC029248.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | NM_020509.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | AF205951.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257951 | AF290872.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133935 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133403 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | AK007553.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | BC029248.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | NM_020509.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | AF205951.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | AF290872.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | AK007553.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | BC029248.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | NM_020509.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | AF205951.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257951 | AF290872.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30742753 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133403 | - | - |
Whole Mouse Genome G4122A | A_52_P254817 | AK007553.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P254817 | BC029248.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P254817 | NM_020509.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P254817 | AF205951.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P254817 | AF290872.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | AK007553.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | BC029248.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | NM_020509.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | AF205951.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | AF290872.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | AK007553.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | BC029248.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | NM_020509.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | AF205951.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P257951 | AF290872.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101923 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133403 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12756045 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12756043 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12756046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30742742 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742766 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742761 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13019687 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742764 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742750 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12756046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742753 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742754 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742745 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12918727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742769 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742770 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742760 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12918724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742767 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742742 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12756037 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742743 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12756043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30742741 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10436097 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10436096 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10436098 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10436099 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449015_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10436095 | - | - |
Mouse Genome 430 2.0 Array | 1449015_at | - | - |
Mouse Genome 430 2.0 Array | 1449015_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57262_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57262_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1600006089.mm.1 | NM_020509.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600006089.mm.1 | NM_020509.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600006089.mm.1 | TC1600000698.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449015_PM_at | - | - |
Mouse Expression Set 430 | 1449015_at | - | - |
Mouse Genome 430A 2.0 Array | 1449015_at | - | - |
Mouse Genome 430A 2.0 Array | 1449015_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449015_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449015_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 925799 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3840138 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3840138 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003840138 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226472 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1226472 | - | - |
mouse-6 v1.1 (Illumina) | scl057262.3_3-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3840138 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl057262.3_3-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840138 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1226472 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226472 | - | - |
