Assay Details
| Gene Symbol: Fabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021272.3 | NP_067247.1 | 2 - 3 | 337 | 87 | |
| GenBank mRNA | AJ870970.1 | - | 2 - 3 | 337 | 87 | |
| AK002955.1 | - | 2 - 3 | 338 | 87 | ||
| AK021271.1 | - | 2 - 3 | 342 | 87 | ||
| BC055280.1 | - | 2 - 3 | 320 | 5710017 | 87 | |
| BC057090.1 | - | 2 - 3 | 323 | 5700428 | 87 | |
| CT010401.1 | - | 2 - 3 | 251 | 87 | ||
| S69799.1 | - | 2 - 3 | 320 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fabp7
Genomic Map