Assay Details
| Gene Symbol: Itgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159564.1 | NP_001153036.1 | 7 - 8 | 930 | 81 | |
| NM_021359.3 | NP_067334.1 | 4 - 5 | 824 | 81 | ||
| GenBank mRNA | AF115376.1 | - | 7 - 8 | 776 | 81 | |
| AK008867.1 | - | 4 - 5 | 824 | 81 | ||
| AK019511.1 | - | 4 - 5 | 687 | 81 | ||
| AK036439.1 | - | 7 - 8 | 931 | 81 | ||
| AK085900.1 | - | 7 - 8 | 971 | 81 | ||
| BC049185.1 | - | 4 - 5 | 793 | 6475423 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb6
Genomic Map