Assay Details
| Gene Symbol: Itgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159564.1 | NP_001153036.1 | 14 - 15 | 2224 | 59 | |
| NM_021359.3 | NP_067334.1 | 11 - 12 | 2118 | 59 | ||
| GenBank mRNA | AF115376.1 | - | 14 - 15 | 2070 | 59 | |
| AK036439.1 | - | 14 - 15 | 2225 | 59 | ||
| AK085900.1 | - | 14 - 15 | 2265 | 59 | ||
| BC049185.1 | - | 11 - 12 | 2087 | 6475423 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb6
Genomic Map