Assay Details
| Gene Symbol: Ghrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286404.1 | NP_001273333.1 | 2 - 3 | 244 | 80 | |
| NM_021488.5 | NP_067463.2 | 3 - 4 | 295 | 80 | ||
| GenBank mRNA | AB035701.1 | - | 2 - 3 | 237 | 80 | |
| AJ243503.1 | - | 2 - 3 | 260 | 80 | ||
| AK008658.1 | - | 3 - 4 | 265 | 80 | ||
| AK008860.1 | - | 3 - 4 | 265 | 80 | ||
| BC132230.1 | - | 2 - 3 | 261 | 40130507 | 80 | |
| BC132232.1 | - | 2 - 3 | 261 | 40130509 | 80 | |
| FJ914266.1 | - | 2 - 3 | 301 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ghrl
Genomic Map