Assay Details
| Gene Symbol: Stard3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021547.3 | NP_067522.1 | 2 - 3 | 420 | 69 | |
| GenBank mRNA | AK041538.1 | - | 2 - 3 | 376 | 69 | |
| AK150166.1 | - | 2 - 3 | 364 | 69 | ||
| AK150254.1 | - | 2 - 3 | 364 | 69 | ||
| AK155124.1 | - | 2 - 3 | 359 | 69 | ||
| AK160480.1 | - | 2 - 3 | 353 | 69 | ||
| BC003313.1 | - | 2 - 3 | 337 | 3155179 | 69 | |
| X82457.1 | - | 2 - 3 | 404 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard3
Genomic Map